missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human R-Spondin 2 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB3266
This item is not returnable.
View return policy
Description
R-Spondin 2 Monoclonal specifically detects R-Spondin 2 in Human samples. It is validated for Neutralization.
Specifikationer
| R-Spondin 2 | |
| Monoclonal | |
| LYOPH | |
| Neutralization 0.4-2 ug/mL | |
| Q6UXX9 | |
| RSPO2 | |
| Mouse myeloma cell line NS0-derived recombinant human R-Spondin 2 Met1-Gly205 Accession # Q6UXX9 | |
| 100 μg | |
| Primary | |
| Detects human R-Spondin 2 in direct ELISAs. | |
| Human | |
| Purified |
| Neutralization | |
| 879709 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cristin 2, CRISTIN2, hRspo2, MGC35555, MGC43342, Roof plate-specific spondin-2, RSPO2, RSpondin 2, R-spondin 2 homolog (Xenopus laevis), R-spondin-2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 340419 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering