missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human mGluR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB48361-100
This item is not returnable.
View return policy
Description
mGluR1 Monoclonal specifically detects mGluR1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| mGluR1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| glutamate receptor, metabotropic 1, GPRC1A, GRM1, GRM1A, metabotropic glutamate receptor 1, mGluR1, MGLUR1A, MGLUR1GPRC1AmGlu1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human mGluR1a, Met1-Ser522, Accession # Q13255 | |
| 100 μg | |
| Primary | |
| Detects human mGluR1a in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 511601 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q13255 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2911 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction