missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LPAR4/LPA4 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB10217U-100UG
This item is not returnable.
View return policy
Description
P2Y9 Monoclonal specifically detects P2Y9 in Human samples. It is validated for Flow Cytometry.
Specifications
| P2Y9 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| G protein-coupled receptor 23, GPCR23, GPR23, GPR23P2Y purinoceptor 9, LPA4LPA-4, lysophosphatidic acid receptor 4, P2RY9, P2RY9G-protein coupled receptor 23, P2Y5-LIKE, P2Y5-like receptor, P2Y9, P2Y9LPA receptor 4, Purinergic receptor 9 | |
| Human embryonic kidney cell line HEK293-derived transfected with human LPAR4/LPA4, Accession # Q99677 | |
| 100 μg | |
| 2846 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 561203 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| Q99677 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human LPAR4/LPA4 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction