missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-10 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB91842-100
This item is not returnable.
View return policy
Description
IL-10 Monoclonal specifically detects IL-10 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| IL-10 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| Human embryonic kidney cell, HEK293-derived human IL-10 protein, Met1-Asn178, Accession # P22301 | |
| 100 μg | |
| Primary | |
| Detects human IL-10 in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 2472A | |
| Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL | |
| P22301 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3586 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction