missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Histamine H3 R Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB10200V-100UG
This item is not returnable.
View return policy
Description
Histamine H3R Monoclonal specifically detects Histamine H3R in Human samples. It is validated for Flow Cytometry.
Specifications
| Histamine H3R | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| GPCR97, GPCR97G protein-coupled receptor 97, G-protein coupled receptor 97, H3R, HH3R, histamine H3 receptor, Histamine H3R, histamine receptor H3, HRH3 | |
| Synthetic peptide containing human Histamine H3 R, Accession # Q9Y5N1 | |
| 100 μg | |
| 11255 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 1008944 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| Q9Y5N1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Histamine H3 R in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction