missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FFAR2/GPR43 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB10082S-100UG
This item is not returnable.
View return policy
Description
FFAR2/GPR43 Monoclonal specifically detects FFAR2/GPR43 in Human samples. It is validated for Flow Cytometry.
Specifications
| FFAR2/GPR43 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| free fatty acid activated receptor 2, free fatty acid receptor 2, G protein-coupled receptor 43, GPR43, GPR43FFA2R, G-protein coupled receptor 43 | |
| Mouse myeloma cell line NS0-derived recombinant human FFAR2/GPR43, Met1-Glu330, Accession # O15552 | |
| 100 μg | |
| 2867 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 1000624 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| O15552 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human FFAR2/GPR43 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction