missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DLEC/CLEC4C/BDCA-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB62991-100
This item is not returnable.
View return policy
Description
DLEC/CLEC4C/BDCA-2 Monoclonal specifically detects DLEC/CLEC4C/BDCA-2 in Human samples. It is validated for Immunohistochemistry.
Specifications
| DLEC/CLEC4C/BDCA-2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| BDCA2, BDCA-2, BDCA2MGC125791, Blood dendritic cell antigen 2, blood dendritic cell antigen 2 protein, CD303, CD303 antigen, CLEC4C, CLECSF11, CLECSF11PRO34150, CLECSF7, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 11, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 7, C-type lectin domain family 4 member C, C-type lectin superfamily member 7, dendritic cell lectin b, Dendritic lectin, DLEC, DLECMGC125792, HECL, HECLMGC125793, MGC125789, PRO34150 | |
| CHO Chinese hamster ovary cell line transfected with human DLEC/CLEC4C/BDCA-2 Phe46-Ile213 Accession # Q8WTT0 | |
| 100 μg | |
| 170482 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 992258 | |
| Immunohistochemistry 1-25μg/mL | |
| Q8WTT0 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human DLEC/CLEC4C/BDCA-2 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction