missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD14 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB11245G-100UG
This item is not returnable.
View return policy
Description
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry
Specifications
| CD14 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2551A | |
| Flow Cytometry | |
| P08571 | |
| Chinese Hamster Ovary cell line, CHO-derived human CD14 protein, Thr20-Cys352, Accession # P08571 | |
| 100 μg | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction