missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Caspr2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5145-100
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Caspr2 Monoclonal specifically detects Caspr2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Tekniske data
| Caspr2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| AUTS15, CASPR2DKFZp781D1846, CDFE, Cell recognition molecule Caspr2, CNTNAP2, contactin associated protein-like 2, contactin-associated protein 2, contactin-associated protein-like 2, homolog of Drosophila neurexin IV, KIAA0868, NRXN4, NRXN4Caspr2, PTHSL1 | |
| Mouse myeloma cell line NS0-derived recombinant human Caspr2, Pro29-Ala1262, Accession # Q9UHC6 | |
| 100 μg | |
| Primary | |
| Detects human Caspr2 in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 840727 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q9UHC6 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 26047 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion