missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Mouse, Alexa Fluor 405, Clone: 1025527, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC42402V-100UG
This item is not returnable.
View return policy
Description
GFP Monoclonal antibody specifically detects GFP in Broad Species Reactivity samples. It is validated for Flow Cytometry
Specifications
| GFP | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Reconstitution default not known | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 1025527 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| eGFP, Enhanced Green Fluorescent Protein, GFPuv, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| E. coli-derived recombinant GFP, aa 2-238 | |
| 100 μg | |
| Primary | |
| Multi-species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction