missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TNF RII/TNFRSF1B PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB11262P-100
This item is not returnable.
View return policy
Description
TNF RII/TNFRSF1B Monoclonal antibody specifically detects TNF RII/TNFRSF1B in Human samples. It is validated for Flow Cytometry
Specifications
| TNF RII/TNFRSF1B | |
| Monoclonal | |
| PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| E. coli-derived recombinant human TNF RIIExtracellular domain | |
| 100 Tests | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 22221 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction