missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NK1R PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB66871P-100
This item is not returnable.
View return policy
Description
NK1R Monoclonal specifically detects NK1R in Human samples. It is validated for Flow Cytometry.
Specifications
| NK1R | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and sodium azide | |
| NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPR, SPRsubstance-P receptor, TAC1R, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor, TACR1 | |
| HEK293 human embryonic kidney cell line transfected with human NK1R, Accession No. P25103 | |
| 100 Tests | |
| 6869.0 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 694501 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P25103 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human NK1R in ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction