missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ 100 μL, Model 1710 TLL SYR, Instrument Syringe

Product Code. 11572475
Change view
Click to view available options
Volume (Metric):
100 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11572475 100 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11572475 Supplier Hamilton™ Supplier No. 988020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The 1700 series syringes are the Gastight version of the original 700 series syringes. This series of syringes is ideal for dispensing volumes from 1 μL up to 500 μL. A variety of different terminations and needle options are available in this series.

Specification Sheet

  • Volume – 100 μL
  • Syringe Type – Gastight
  • Syringe Series – 1700
  • Termination – PTFE Luer Lock (TLL)
  • Needle – Instrument Syringe
  • Gauge – No Needle Included
  • Point Style – No Needle Included
  • Needle Length – No Needle Included
  • Plunger Style – Side Post Mounting (XL)
  • Fluid Path – Borosilicate glass, PTFE
  • Autoclavable – No
  • Gas Sterilizable – Yes
  • Pressure – 1000 psig (68.9 bar)
  • Maximum Temperature – 115°C / 239°F
  • Minimum Temperature – 10°C / 50°F
  • Barrel Inner Diameter – 0.057 inches (1.46 mm)
  • Barrel Outer Diameter – 0.305 inches (7.75 mm)
  • Std Needle Dead Volume – N/A
  • Sales Quantity – 1 syringe per pack

Specifications

Product Type Model 1710 TLL SYR, Instrument Syringe
Volume (Metric) 100 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.