missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ 100 Base-Pair DNA Ladder, Express

Product Code. 16022575
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16022575 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16022575 Supplier Abnova™ Supplier No. D0028.500uL
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Consists of multiple repeats of 100-bp fragments from 100 bp up to 1000 bp.

  • Supplied in sufficient quantity for 100 loadings using 4-5 ul per loading.
  • Not designed for quantitating DNA concentration in a sample.
  • Ladders available premixed as ready-to-use format premixed with loading-dye-solution (Express).

Specifications

For Use With (Application) Electrophoresis
Quantity 500 μL
Storage Requirements Store at -20°C Stable (at room temperature for 6 months)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.