missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ 10 x Taq Buffer

Product Code. 16002575
Change view
Click to view available options
Quantity:
1.8 mL
Unit Size:
1.8mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16002575 1.8 mL 1.8mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16002575 Supplier Abnova™ Supplier No. D0024.1.8mL
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

10 x Taq Buffer is a ready-to-use buffer to be used for Taq, Hot Taq, Red Taq and Long Taq enzymes in PCR reactions. Does not contain MgCl2.

Specifications

For Use With (Application) Standard PCR
Quantity 1.8 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.