missing translation for 'onlineSavingsMsg'
Learn More

10-Undecynoic acid, 96%

Product Code. 11444894
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444894 25 g 25g
11434894 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444894 Supplier Thermo Scientific Alfa Aesar Supplier No. A13815.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2777-65-3 | C11H18O2 | 182.263 g/mol

10-Undecynoic acid may be employed as a biochemical probe in an assay for the microsomal hydroxylation of lauric acid (LA), based on HPLC with flow-through radiochemical detection. It was employed as model compound to investigate the microwave assisted surface click reactions catalyzed with Cu(II)/sodium L-ascorbate. It is reported as highly selective irreversible inhibitor of hepatic ω- and ω-1-lauric acid hydroxylases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10-Undecynoic acid may be employed as a biochemical probe in an assay for the microsomal hydroxylation of lauric acid (LA), based on HPLC with flow-through radiochemical detection. It was employed as model compound to investigate the microwave assisted surface click reactions catalyzed with Cu(II)/sodium L-ascorbate. It is reported as highly selective irreversible inhibitor of hepatic ω- and ω-1-lauric acid hydroxylases.

Solubility
Sparingly Soluble in water.

Notes
Store at 4°C. Store away from incompatible materials such as oxidizing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2777-65-3
Molecular Formula C11H18O2
Molecular Weight (g/mol) 182.263
MDL Number MFCD00014389
InChI Key OAOUTNMJEFWJPO-UHFFFAOYSA-N
Synonym 10-undecynoic acid, hendecynoic acid, unii-2f79g7h1wy, undec-10-ynoicacid, acmc-1cp9s, undec-1-yn-11-oic acid, 4-02-00-01738 beilstein handbook reference
PubChem CID 31039
IUPAC Name undec-10-ynoic acid
SMILES C#CCCCCCCCCC(=O)O

Specifications

Melting Point 40°C to 43°C
Boiling Point 174°C to 176°C (4 mmHg)
Flash Point 161°C (321°F)
Odor Odorless
Quantity 25 g
Beilstein 1704918
Solubility Information Sparingly Soluble in water.
Formula Weight 182.26
Percent Purity 96%
Chemical Name or Material 10-Undecynoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.