Learn More
10-Undecynoic acid, 96%
Description
10-Undecynoic acid may be employed as a biochemical probe in an assay for the microsomal hydroxylation of lauric acid (LA), based on HPLC with flow-through radiochemical detection. It was employed as model compound to investigate the microwave assisted surface click reactions catalyzed with Cu(II)/sodium L-ascorbate. It is reported as highly selective irreversible inhibitor of hepatic ω- and ω-1-lauric acid hydroxylases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
10-Undecynoic acid may be employed as a biochemical probe in an assay for the microsomal hydroxylation of lauric acid (LA), based on HPLC with flow-through radiochemical detection. It was employed as model compound to investigate the microwave assisted surface click reactions catalyzed with Cu(II)/sodium L-ascorbate. It is reported as highly selective irreversible inhibitor of hepatic ω- and ω-1-lauric acid hydroxylases.
Solubility
Sparingly Soluble in water.
Notes
Store at 4°C. Store away from incompatible materials such as oxidizing agents
Specifications
Specifications
| Melting Point | 40°C to 43°C |
| Boiling Point | 174°C to 176°C (4 mmHg) |
| Flash Point | 161°C (321°F) |
| Odor | Odorless |
| Quantity | 5 g |
| Beilstein | 1704918 |
| Solubility Information | Sparingly Soluble in water. |
| Formula Weight | 182.26 |
| Percent Purity | 96% |
| Chemical Name or Material | 10-Undecynoic acid |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.