missing translation for 'onlineSavingsMsg'
Learn More

10-Undecyn-1-ol, 96%

Product Code. 11362627
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11362627

Brand: Thermo Scientific Alfa Aesar L11807.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2774-84-7 | C11H20O | 168.28 g/mol

10-Undecyn-1-ol exhibits an antifungal activity. It is also used in the esterification of pentanoic and stearic acids in the presence of lipase. It is also employed as a reagent in the preparation of 11-bromo-undec-1-yne by reacting with carbon tetrabromide and triphenylphosphine. Further, it acts as an intermediate in synthetic chemistry. In addition to this, it serves as a precursor in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10-Undecyn-1-ol exhibits an antifungal activity. It is also used in the esterification of pentanoic and stearic acids in the presence of lipase. It is also employed as a reagent in the preparation of 11-bromo-undec-1-yne by reacting with carbon tetrabromide and triphenylphosphine. Further, it acts as an intermediate in synthetic chemistry. In addition to this, it serves as a precursor in organic synthesis.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2774-84-7
Molecular Formula C11H20O
Molecular Weight (g/mol) 168.28
MDL Number MFCD00041675
InChI Key YUQZOUNRPZBQJK-UHFFFAOYSA-N
Synonym 10-undecyn-1-ol, 10-undecynol, 11-hydroxyundecyne, 1-undecyn-11-ol, acmc-1cmhp, 11-hydroxy-1-undecyne, 10-undecyn-1-ol,, 10-undecyn-1-ol gc
PubChem CID 76015
IUPAC Name undec-10-yn-1-ol
SMILES C#CCCCCCCCCCO

Specifications

Density 0.873
Boiling Point 110°C to 111°C (4 mmHg)
Refractive Index 1.4565
Quantity 1 g
Beilstein 1702764
Solubility Information Immiscible with water.
Formula Weight 168.28
Percent Purity 96%
Chemical Name or Material 10-Undecyn-1-ol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt