missing translation for 'onlineSavingsMsg'
Learn More

10-Undecenoic Acid, TRC

Product Code. 30455541
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455541 100 g 100g
30452632 250 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30455541 Supplier Toronto Research Chemicals Supplier No. TRCU788820100G
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-38-9
Molecular Formula C11 H20 O2
Molecular Weight (g/mol) 184.28
Synonym Undecylenic acid, Undec-10-enoic acid
IUPAC Name undec-10-enoic acid
SMILES OC(=O)CCCCCCCCC=C

Specifications

Quantity 100 g
InChI Formula InChI=1S/C11H20O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2H,1,3-10H2,(H,12,13)
Formula Weight 184.1463
Chemical Name or Material Undecylenic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.