missing translation for 'onlineSavingsMsg'
Learn More

10-Undecenoic acid, MedChemExpress

Product Code. 16458506
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458506 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458506 Supplier Medchem Express Supplier No. HYB0914
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 10-Undecenoic acid was used as a starting reagent in the syntheses of Pheromone (11Z)-hexadecenal.

10-Undecenoic acid was used as a starting reagent in the syntheses of Pheromone (11Z)-hexadecenal.

Specifications

Chemical Name or Material 10-Undecenoic acid
Physical Form Liquid
CAS 112-38-9
Color Colorless
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C11H20O2
Synonym Undecylenic acid
Solubility Information DMSO : ≥ 50 mg/mL (271.33 mM)
SMILES C=CCCCCCCCCC(O)=O
Molecular Weight (g/mol) 184.28
Formula Weight 184.28
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.