missing translation for 'onlineSavingsMsg'
Learn More

10-Undecen-1-ol, 99%

Product Code. 11467856
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467856 25 g 25g
11477856 100 g 100g
11487856 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467856 Supplier Thermo Scientific Alfa Aesar Supplier No. A14002.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-43-6 | C11H22O | 170.296 g/mol

10-Undecen-1-ol is used as a flavoring agent. It reacts with tosylated beta-cyclodextrin to prepare beta-cyclodextrin undecenylether. It is used in the synthesis of 10-Undecenylacrylat by reacting with acryloyl and triethylamine. It reacts with carbon monoxide in the presence of dicobalt octacarbonyl and pyridine as a base at pressures greater than 150 bar and temperatures in the range of 100 to 2000C to linear and branched polyesters having relatively high molecular weights (Mn> 10,000 g / mol) in good yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10-Undecen-1-ol is used as a flavoring agent. It reacts with tosylated beta-cyclodextrin to prepare beta-cyclodextrin undecenylether. It is used in the synthesis of 10-Undecenylacrylat by reacting with acryloyl and triethylamine. It reacts with carbon monoxide in the presence of dicobalt octacarbonyl and pyridine as a base at pressures greater than 150 bar and temperatures in the range of 100 to 2000C to linear and branched polyesters having relatively high molecular weights (Mn> 10,000 g / mol) in good yields.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-43-6
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.296
MDL Number MFCD00004750
InChI Key GIEMHYCMBGELGY-UHFFFAOYSA-N
Synonym 10-undecen-1-ol, undecylenic alcohol, undecylenyl alcohol, 10-undecenol, 1-undecen-11-ol, 10-undecylen-1-ol, 11-hydroxy-1-undecene, omega-undecenyl alcohol, undecen-1-ol, .omega.-undecenyl alcohol
PubChem CID 8185
IUPAC Name undec-10-en-1-ol
SMILES C=CCCCCCCCCCO

Specifications

Melting Point -3°C
Density 0.848
Boiling Point 132°C to 133°C (15 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.45
Quantity 25 g
Beilstein 1753225
Solubility Information Miscible with alcohol and ether. Immiscible with water.
Formula Weight 170.3
Percent Purity 99%
Chemical Name or Material 10-Undecen-1-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.