missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IFN-alpha-2B Protein
Click to view available options
Quantity:
10 μg
50 μg
Packungsgröße:
10µg
50µg
Beschreibung
Measured in anti-viral assays using HeLa human cervical epithelial carcinoma cells infected with encephalomyocarditis (EMC) virus. Meager, A. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 129. The ED50 fot this effect is 3.00-60.0 pg/mL.
Spezifikation
Spezifikation
| Accession Number | NP_000596.2 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 3440 |
| Molecular Weight (g/mol) | M.W.-observed: 18-22 kDa, under reducing conditions., M.W.-theroretical: 19 kDa |
| Quantity | 10 μg |
| Source | Human embryonic kidney cell, HEK293-derived human IFN-alpha protein Cys24-Glu188 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur