missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human SLC7A2 (NP_001008539.3) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Specifications
Specifications
| Gene | SLC7A2 |
| Accession Number | NP_001008539.3 |
| Gene ID (Entrez) | 6542 |
| Gene Alias | CAT2, CAT-2, cationic 2, cationic amino acid transporter, y+ system, low affinity cationic amino acid transporter 2, low-affinity cationic amino acid transporter-2, solute carrier family 7 (cationic amino acid transporter, y+ system), member 2, Solute carrier family 7 member 2 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction