missing translation for 'onlineSavingsMsg'
Learn More

10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane, 95%

Codice prodotto. 15478785
Cambia vista
Click to view available options
Quantity:
250 mg
1 g
Dimensione della confezione:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
15478785 250 mg 250mg
15468785 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15478785 Fornitore Thermo Scientific Alfa Aesar N. del fornitore H34117.MD
 Restricted Item
Estimated Shipment: 07-12-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 944721-52-2 | C20H29F5O5Si | 472.524 g/mol

It finds its application in optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in optimized immobilization of lectins using self-assembled monolayers on polysilicon encoded materials for cell tagging.

Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly sealed and place it in a cool, dry and well ventilated place. Store under inert gas.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 944721-52-2
Molecular Formula C20H29F5O5Si
Molecular Weight (g/mol) 472.524
MDL Number MFCD13181915
InChI Key KSYPNQIBGIOYPZ-UHFFFAOYSA-N
Synonym 11-pentafluorophenylundecanoato trimethoxysilane, 10-pentafluorophenoxycarbonyl decyltrimethoxysilane, 2,3,4,5,6-pentafluorophenyl 11-trimethoxysilyl undecanoate
PubChem CID 23582157
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 11-trimethoxysilylundecanoate
SMILES CO[Si](CCCCCCCCCCC(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC

Specifica

Boiling Point 145°C to 150°C (0.05mmHg)
Quantity 250 mg
Refractive Index 1.4370-1.4410
Sensitivity Moisture sensitive
Formula Weight 472.52
Percent Purity 95%
Chemical Name or Material 10-(Pentafluorophenoxycarbonyl)decyltrimethoxysilane

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.