missing translation for 'onlineSavingsMsg'
Learn More

10-Oxo Docetaxel, TRC

Product Code. 17979749
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
1mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17979749 1 mg 1mg
18080230 25 mg 25mg
18081740 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17979749 Supplier Toronto Research Chemicals Supplier No. TRCO8558001MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167074-97-7
Molecular Formula C43 H51 N O14
Molecular Weight (g/mol) 805.86
Synonym Benzenepropanoic acid, β-[[(1,1-dimethylethoxy)carbonyl]amino]-α-hydroxy-, (2aR,4S,4aS,9S,11S,12S,12aR,12bS)-12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5,6-dioxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)-, Benzenepropanoic acid, β-[[(1,1-dimethylethoxy)carbonyl]amino]-α-hydroxy-, 12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5,6-dioxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, [2aR-[2aα,4β,4aβ,9α(αR*,βS*),11α,12α,12aα,12bα]]-, 5β,20-Epoxy-1,7β-dihydroxy-9,10-dioxotax-11-ene-2α,4,13α-triyl 4-acetate 2-benzoate 13-[(2R,3S)-3-[[(1,1-dimethylethoxy)carbonyl]amino]-2-hydroxy-3-phenylpropanoate, 10-Deoxy-10-oxodocetaxel, Docetaxel Trihydrate Imp B (EP), 6-Oxodocetaxel, (2aR,4S,4aS,9S,11S,12S,12aR,12bS)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-Dodecahydro-4,9,11,12,12b-pentahydroxy-4a,8,13,13-tetramethyl-7,11-methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5,6-dione 12b-acetate 12-benzoate 9-ester with (2R,3S)-N-tert-butoxycarbonyl-3-phenylisoserine
IUPAC Name [(1S,2S,3R,4S,7R,9S,10S,15S)-4-acetyloxy-1,9-dihydroxy-15-[(2R,3S)-2-hydroxy-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoyl]oxy-10,14,17,17-tetramethyl-11,12-dioxo-6-oxatetracyclo[11.3.1.03,10.04,7]heptadec-13-en-2-yl] benzoate
SMILES CC(=O)O[C@@]12CO[C@@H]1C[C@H](O)[C@]3(C)[C@@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](O)[C@@H](NC(=O)OC(C)(C)C)c6ccccc6)C(=C(C(=O)C3=O)C5(C)C)C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C43H51NO14/c1-22-26(55-37(51)32(48)30(24-15-11-9-12-16-24)44-38(52)58-39(3,4)5)20-43(53)35(56-36(50)25-17-13-10-14-18-25)33-41(8,34(49)31(47)29(22)40(43,6)7)27(46)19-28-42(33,21-54-28)57-23(2)45/h9-18,26-28,30,32-33,35,46,48,53H,19-21H2,1-8H3,(H,44,52)/t26-,27-,28+,30-,32+,33-,35-,41+,42-,43+/m0/s1
Formula Weight 805.331
Percent Purity >95%
Chemical Name or Material 10-Oxo docetaxel
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.