missing translation for 'onlineSavingsMsg'
Learn More

10-Oxo-10,11-Dihydro-5H-dibenz[b,f]azepine, TRC

Product Code. 30137052
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137052 10 mg 10mg
30137922 5 mg 5mg
30007445 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137052 Supplier Toronto Research Chemicals Supplier No. TRCO87000010MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21737-58-6
Molecular Formula C14 H11 N O
Molecular Weight (g/mol) 209.24
Synonym 5,11-Dihydro-10H-dibenz[b,f]azepin-10-one
IUPAC Name 6,11-dihydrobenzo[b][1]benzazepin-5-one
SMILES O=C1Cc2ccccc2Nc3ccccc13

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H11NO/c16-14-9-10-5-1-3-7-12(10)15-13-8-4-2-6-11(13)14/h1-8,15H,9H2
Formula Weight 209.0841
Chemical Name or Material 10-Oxo-10,11-Dihydro-5H-dibenz[b,f]azepine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.