missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ WS 383

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
WS 383 is a potent and reversible DCN1-UBC12 interaction inhibitor (IC50 = 11 nM). The compound displays selectivity for DCN1-UBC12 over inhibition of a panel of kinases. including Bruton's tyrosine kinase (BTK), cyclin-dependent kinases (CDKs) and epidermal growth factor receptor (EGFR). In vitro, WS 383 selectively inhibits cullin-3 neddylation over other cullins, and induces accumulation of p21, p27 and NRF2.
Specifications
Specifications
| Chemical Name or Material | 5-[[2-[[(4-Chlorophenyl)methyl]thio]-5-methyl[1,2,4 ]triazolo[1,5-a]pyrimidin-7-yl]thio]-N,N-dimethyl-1 H-tetrazole-1-ethanamine hydrochloride |
| CAS | 2247544-02-9 |
| Quantity | 10 mg |
| Target | Ubiquitin Ligase (E3) Inhibitors |
| Molecular Formula | C18H20ClN9S2.HCl |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction