missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ UBP 302
GREENER_CHOICE

Product Code. 30001294
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001294

Brand: Tocris Bioscience™ 2079/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective kainate antagonist; active enantiomer of UBP 296 (Cat. No. 2078)

Potent and selective GluK1 (formally GluR5)-subunit containing kainate receptor antagonist (apparent KD = 402 nM); active enantiomer of UBP 296 (Cat. No. 2078). Displays ∽ 260-fold selectivity over AMPA receptors, ∽ 90-fold selectivity over recombinant human GluK2 (formally GluR6) and GluK5 (formally KA2)-containing kainate receptors and has little or no action at NMDA or group I mGlu receptors. Selectively blocks kainate receptor-mediated LTP induction in rat hippocampal mossy fibers. Also protects against soman-induced seizures and neuronal degeneration for up to 30 days when administered one hour after soman exposure.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 745055-91-8
Molecular Formula C15H15N3O6

Specifications

Quantity 10 mg
Formula Weight 333.3
Purity >98%
Molecular Weight (g/mol) 333.3
Chemical Name or Material UBP 302
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.