missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SPRi 3

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SPRi 3 is a potent sepiapterin reductase (SPR) inhibitor (IC50 = 53 - 74 nM for human SPR). Inhibits BH4 synthesis pathway and attenuates proliferation in naive CD4+ T cells. Suppresses proliferation of human effector CD4+ T cells following stimulation. Reduces SPR activity in mouse primary cultures of sensory neurons (IC50 = 450 nM). Ameliorates colitis, suppressing the intestinal infiltration of T cells and other immune cells in vivo. Also reduces immune-cell infiltration into mouse lungs. Reduces neuropathic and inflammatory pain in mouse models.
Specifications
Specifications
| Chemical Name or Material | N-[2-(5-Hydroxy-2-methyl-1H-indol-3-yl)ethyl]-2-methoxyacetamide |
| CAS | 1292285-54-1 |
| Quantity | 10 mg |
| Molecular Formula | C14H18N2O3 |
| Formula Weight | 262.3 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction