missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PSB CB5
GREENER_CHOICE

Product Code. 18779106
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18779106

Brand: Tocris Bioscience™ 6372/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective GPR18 antagonist

PSB CB5 is a selective GPR18 antagonist (IC50 = 0.279 μM). Exhibits >36-fold selectivity versus CB1 and GPR55 and 14-fold selectivity versus CB2 receptors. Inhibits N-Arachidonylglycine (Cat. No. 1445) induced acrosomal exocytosis in human spermatozoa.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2 Z)-2 [[3-[(4-Chlorophenyl)methoxy]phenyl]methylene]-6,7-dihydro-5 H-imidazo[2,1-b][1,3 ]thiazin-3(2 H)-one
CAS 1627710-30-8
Quantity 10 mg
Target Additional Cannabinoid-related Compounds
Molecular Formula C20H17ClN2O2S
Formula Weight 2020
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.