missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NNC 55-0396 dihydrochloride
GREENER_CHOICE

Product Code. 30001337
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30001337

Brand: Tocris Bioscience™ 2268/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly selective CaV3.x blocker

Highly selective T-type calcium channel blocker. Displays IC50 values of 6.8 and > 100 μM for inhibition of Cav3.1 T-type channels and HVA currents respectively in INS-1 cells.
TRUSTED_SUSTAINABILITY

Specifications

CAS 357400-13-6
Quantity 10 mg
Molecular Formula C30H40Cl2FN3O2
Formula Weight 564.57
Purity >98%
Molecular Weight (g/mol) 564.567
Chemical Name or Material NNC 55-0396 dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.