missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Istradefylline
GREENER_CHOICE

Product Code. 30002035
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30002035

Brand: Tocris Bioscience™ 5147/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective adenosine A2A receptor antagonist

Potent and selective adenosine A2A receptor antagonist (Ki values are 2.2 and 150 nM for A2A and A1 receptors respectively). Anticataleptic and antiparkinson agent; reverses drug-induced motor dysfunction in animal models.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155270-99-8
Molecular Formula C20H24N4O4

Specifications

Quantity 10 mg
Formula Weight 384.43
Physical Form Solid
Color White
Purity >99%
Molecular Weight (g/mol) 384.44
Chemical Name or Material Istradefylline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.