missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Cilengitide

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Potent and selective inhibitor of integrins αvβ3 and αvβ5 (IC50 values are 4.1 and 70 nM, respectively). Exhibits ∽10-fold selectivity over gpIIb/IIIa. Increases endothelial monolayer permeability. Also exhibits antiangiogenic activity.
Specifications
Specifications
| Purity | 95% |
| CAS | 188968-51-6 |
| Content And Storage | Store at −20°C |
| Molecular Weight (g/mol) | 588.666 |
| Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction