missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 10 μL Microvolume Filter Tips

Product Code. 12633706 Shop All Axygen Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Universal Fit Micro
Tip Style:
Maxymum Recovery, Universal
Microvolume
Universal, Maxymum Recovery
Universal, Microvolume
Format:
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 10000
Pack of 4800
Pack of 960
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Tip Style Format unitSize
12633706 Universal Fit Micro Universal, Maxymum Recovery Rack Pack of 960
12796739 Universal Fit Micro Universal, Maxymum Recovery Rack Pack of 4800
12106890 Universal Fit Micro Universal, Microvolume Bulk Pack of 1000
12511358 Universal Fit Micro Universal, Microvolume Bulk Pack of 10000
11346074 Universal Fit Micro Universal, Microvolume Rack Pack of 960
12706749 Universal Fit Micro Universal, Microvolume Rack Pack of 4800
12756749 Universal Fit Micro Maxymum Recovery, Universal Rack Pack of 4800
12531358 Universal Fit Micro Microvolume Bulk Pack of 10000
12116890 Universal Fit Micro Microvolume Bulk Pack of 1000
12766749 Universal Fit Micro Microvolume Rack Pack of 4800
12555537 Universal Fit Micro Microvolume Rack Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 12633706 Supplier Axygen™ Supplier No. TF300LRS
Add to basket
Edge
Add to basket

Axygen Universal Tips designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.

  • Technologically advanced filter material allows free passage of air while blocking aerosol contamination.
  • Filter membrane contains no leachable additives.
  • Axygen™ Maxymum Recovery™ surface improves accuracy and ensures maximum sample recovery.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Human gDNA- and PCR-inhibition-free
  • Order Info

    • Non-Filtered bulk: 1000 tips/pack, 240 packs/case, Filtered bulk: 1000 tips/pack, 10 packs/case
    • Racked: 96 tips/rack, 10 racks/pack, 5 packs/case
    • Stack Racks: 96 tips/rack, 5 racks/stack,10 stacks/case

    TRUSTED_SUSTAINABILITY

    Specifications

    Volume (Metric) 0.5 to 10 μL
    Volume (Metric) Dispensing 0.5 to 10 μL
    Filtered Filtered
    Sterility Sterile
    Color Clear
    Autoclavable Not Autoclavable
    Certifications/Compliance USP 3127
    Clean Claims DNase, Rnase Free
    For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
    Format Rack
    Tip Style Universal, Maxymum Recovery
    Packaging Rack
    Quantity 960/Pk.
    Product Type Filtered Pipette Tips
    Includes 10 Racks x 96 Tips
    Green Features Bulk Handheld Tips Use on Average89% Less Resin Than Their Pre-racked Alternatives.
    Disposable Single-use
    For Use With (Equipment) Universal Fit Micro
    Graduated No
    Material Polypropylene
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.