Learn More
Axygen™ 10 μL Microvolume Filter Tips
Axygen Universal Tips designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
Brand:  Axygen™ TF-400
76.51 GBP valid until 2024-12-31
Use promo code "23272" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.20700kg
 View more versions of this product
Product Code. 12116890
  • £86.95 / Pack of 1000
Estimated Shipment: 06-11-2024
to see stock.
Add to basket
Description
Description
  • Technologically advanced filter material allows free passage of air while blocking aerosol contamination.
  • Filter membrane contains no leachable additives.
  • Axygen™ Maxymum Recovery™ surface improves accuracy and ensures maximum sample recovery.
  • RNase-/DNase-free
  • Nonpyrogenic
  • Human gDNA- and PCR-inhibition-free
  • Specifications
    Specifications
    0.5 to 10 μL
    Non-sterile
    USP 3127
    Allows recovery of costly samples with improved precision and accuracy
    Microvolume
    Eppendorf™ Research™ pipettes, 0.5 to 10μL
    Filtered
    Clear
    DNase, Rnase Free
    Bulk
    1,000 Tips/Pk., 10 Packs/Cs.
    Polypropylene