Learn More
Hamilton™ 10 μL 1700 Series Gas Tight Syringe
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
Brand:  Hamilton™ CAL80075
 View more versions of this product
Product Code. 17834412
  • £234.26 / Each
Explore more special offers
Description
Description
  • This series of syringes are ideal for dispensing volumes up to 10 μL.
  • A variety of different terminations and needle options are available in this series.
Specifications
Specifications
Not Autoclavable
26s G
0.005 in.
0.0187 in.
0.460 mm
0.47 mm
Gas Tight Syringe
50°C
1701 N Syringe
1
51 mm
Cemented
Stainless Steel
0.018 in.
0.13 mm
0.260 in.
6.60 mm
2 in.
122°F
50°F
10°C
10 μL
Cone