missing translation for 'onlineSavingsMsg'
Learn More

10-Hydroxygeraniol, TRC

Product Code. 30397243
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397243 250 mg -
30393210 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397243 Supplier Toronto Research Chemicals Supplier No. TRCH939740250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26488-97-1
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
Synonym (E,E)-2,6-Dimethyl-2,6-octadiene-1,8-diol, (E,E)-2,6-Dimethylocta-2,6-diene-1,8-diol
IUPAC Name (2E,6Z)-2,6-dimethylocta-2,6-diene-1,8-diol
SMILES OC/C(C)=C/CC/C(C)=C\CO

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H18O2/c1-9(6-7-11)4-3-5-10(2)8-12/h5-6,11-12H,3-4,7-8H2,1-2H3/b9-6-,10-5+
Formula Weight 170.1307
Chemical Name or Material 10-Hydroxygeraniol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.