missing translation for 'onlineSavingsMsg'
Learn More

10-Hydroxydecanoic acid, 96%

Product Code. 15457238
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457238 5 g 5g
15447238 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457238 Supplier Thermo Scientific Alfa Aesar Supplier No. H60743.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1679-53-4 | C10H20O3 | 188.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1679-53-4
Molecular Formula C10H20O3
Molecular Weight (g/mol) 188.27
MDL Number MFCD00010510
InChI Key YJCJVMMDTBEITC-UHFFFAOYSA-N
Synonym 10-hydroxydecanoate, decanoic acid, 10-hydroxy, unii-np03xo416b, 10-hydroxy capric acid, 10-hydroxy-decanoic acid, 1-hydroxydecanoicacid, 10-oh-capric acid, 10-hydroxycapric acid, 10-oh-decanoic acid, 10-hydroxydecanoicacid
PubChem CID 74300
ChEBI CHEBI:17409
IUPAC Name 10-hydroxydecanoic acid
SMILES OCCCCCCCCCC(O)=O

Specifications

Melting Point 75°C to 77°C
Linear Formula HO(CH2)9CO2H
Quantity 5 g
Formula Weight 188.26
Percent Purity 96%
Chemical Name or Material 10-Hydroxydecanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.