missing translation for 'onlineSavingsMsg'
Learn More

10-Hydroxy Nortriptyline Maleate, TRC

Product Code. 30399462
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
25mg
Product Code. Quantity unitSize
30399462 25 mg 25mg
30389534 2.5 mg 2.50mg
2 options
This item is not returnable. View return policy

Product Code. 30399462

Brand: Toronto Research Chemicals TRCH94882025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74853-74-0
Molecular Formula C23H25NO
Molecular Weight (g/mol) 395.45
Synonym (5E)-10,11-Dihydro-5-[3-(methylamino)propylidene]-5H-dibenzo[a,d]cyclohepten-10-ol (2Z)-2-Butenedioate, (E)-10-Hydroxynortriptyline Maleate, 10-Hydroxy-(E)-nortriptyline Maleate, trans-10-Hydroxynortriptyline Maleate
SMILES CNCC/C=C/1\C2=CC=CC=C2CC(C3=CC=CC=C13)O.C(=C/C(=O)O)/C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H21NO/c1-20-12-6-11-16-15-8-3-2-7-14(15)13-19(21)18-10-5-4-9-17(16)18/h2-5,7-11,19-21H,6,12-13H2,1H3/b16-11+
Formula Weight 395.1733
Chemical Name or Material 10-Hydroxy Nortriptyline Maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.