missing translation for 'onlineSavingsMsg'
Learn More

10-Hydroxy Camptothecin, TRC

Product Code. 30392146
Change view
Click to view available options
Quantity:
1 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392146 10 mg
30395039 100 mg
30408710 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392146 Supplier Toronto Research Chemicals Supplier No. TRCH87500010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19685-09-7
Molecular Formula C20 H16 N2 O5
Molecular Weight (g/mol) 364.35
Synonym 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 4-ethyl-4,9-dihydroxy-, (4S)-, 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 4-ethyl-4,9-dihydroxy-, (S)-, Camptothecine, 10-hydroxy- (8CI), (+)-10-Hydroxycamptothecin, (S)-10-Hydroxycamptothecin, 10-Hydroxycamptothecin, 10-Hydroxycamptothecine, ChEMBL 273862, Hydroxycamptothecin, NSC 107124
IUPAC Name (4S)-4-Ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
SMILES CC[C@@]1(O)C(=O)OCC2=C1C=C3N(Cc4cc5cc(O)ccc5nc34)C2=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H16N2O5/c1-2-20(26)14-7-16-17-11(5-10-6-12(23)3-4-15(10)21-17)8-22(16)18(24)13(14)9-27-19(20)25/h3-7,23,26H,2,8-9H2,1H3/t20-/m0/s1
Formula Weight 364.1059
Percent Purity >95%
Chemical Name or Material (S)-10-Hydroxycamptothecin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.