missing translation for 'onlineSavingsMsg'
Learn More

10-Hydroxy Camptothecin 9,9’-Methylene Dimer, TRC

Product Code. 30391079
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391079 25 mg 25mg
30404711 1 mg 1mg
30395183 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391079 Supplier Toronto Research Chemicals Supplier No. TRCH87500525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10-Hydroxy Camptothecin 9,9’-Methylene Dimer
Analyte or Component Names 10-Hydroxy Camptothecin 9,9’-Methylene Dimer
Shipping Condition Room Temperature
Molecular Formula C41H32N4O10
InChI Formula InChI=1S/C41H32N4O10/c1-3-40(52)26-12-30-34-18(14-44(30)36(48)24(26)16-54-38(40)50)9-20-22(32(46)7-5-28(20)42-34)11-23-21-10-19-15-45-31(35(19)43-29(21)6-8-33(23)47)13-27-25(37(45)49)17-55-39(51)41(27,53)4-2/h5-10,12-13,46-47,52-53H,3-4,11,14-17H2,1-2H3/t40-,41-/m0/s1
Solution Type Neat
SMILES O=C(OC1)[C@@](CC)(O)C2=C1C(N(C3)C(C(C3=C4)=NC5=C4C(CC6=C(O)C=CC7=C6C=C(CN8C9=CC%10=C(COC([C@]%10(O)CC)=O)C8=O)C9=N7)=C(O)C=C5)=C2)=O
IUPAC Name (4S,4'S)-10,10'-methylenebis(4-ethyl-4,9-dihydroxy-1,12-dihydro-14H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H)-dione)
Industry Type Pharmaceutical
Molecular Weight (g/mol) 740.71
Quantity 25 mg
Formula Weight 740.21
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.