missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-Formyl Folic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Formylfolic acid |
| CAS | 134-05-4 |
| Molecular Formula | C20 H19 N7 O7 |
| Quantity | 50 mg |
| Synonym | N-[4-[[(2-Amino-3,4-dihydro-4-oxo-6-pteridinyl)methyl]formylamino]benzoyl]-L-glutamic acid, Glutamic acid, N-[p-[N-[(2-amino-4-hydroxy-6-pteridinyl)methyl]formamido]benzoyl]- (6CI), Glutamic acid, N-[p-[N-[(2-amino-4-hydroxy-6-pteridinyl)methyl]formamido]benzoyl]-, L- (7CI,8CI), L-Glutamic acid, N-[4-[[(2-amino-1,4-dihydro-4-oxo-6-pteridinyl)methyl]formylamino]benzoyl]- (9CI), N-[4-[[(2-Amino-3,4-dihydro-4-oxo-6-pteridinyl)methyl]formylamino]benzoyl]-L-glutamic acid, 10-Formylfolic acid, 10-Formylpteroylglutamic acid, N10-Formylfolic acid |
| InChI Formula | InChI=1S/C20H19N7O7/c21-20-25-16-15(18(32)26-20)23-11(7-22-16)8-27(9-28)12-3-1-10(2-4-12)17(31)24-13(19(33)34)5-6-14(29)30/h1-4,7,9,13H,5-6,8H2,(H,24,31)(H,29,30)(H,33,34)(H3,21,22,25,26,32)/t13-/m0/s1 |
| SMILES | NC1=NC(=O)c2nc(CN(C=O)c3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O)cnc2N1 |
| IUPAC Name | (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methyl-formylamino]benzoyl]amino]pentanedioic acid |
| Molecular Weight (g/mol) | 469.41 |
| Formula Weight | 469.1346 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?