missing translation for 'onlineSavingsMsg'
Learn More

10-Deoxy-10-oxo-7-epi-docetaxel, Mikromol

Product Code. 30461326
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461326 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461326 Supplier Mikromol Supplier No. MM3396.080025
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

10-Deoxy-10-oxo-7-epi-docetaxel, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Epi-10-oxo-docetaxel (Docetaxel Impurity D)
CAS 162784-72-7
Analyte or Component Names 7-Epi-10-oxo-docetaxel (Docetaxel Impurity D)
Shipping Condition Ice pack (-20°C)
Molecular Formula C43 H51 N O14
Certifications/Compliance ISO 9001
Synonym Benzenepropanoic acid, β-[[(1,1-dimethylethoxy)carbonyl]amino]-α-hydroxy-, (2aR,4R,4aS,9S,11S,12S,12aR,12bS)-12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5,6-dioxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)-, Benzenepropanoic acid, β-[[(1,1-dimethylethoxy)carbonyl]amino]-α-hydroxy-, 12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5,6-dioxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, [2aR-[2aα,4α,4aβ,9α(αR*,βS*),11α,12α,12aα,12bα]]-, 4-epi-6-Oxodocetaxel, 5β,20-Epoxy-1,7α-dihydroxy-9,10-dioxotax-11-ene-2α,4,13α-triyl 4-acetate 2-benzoate 13-[(2R,3S)-3-[[(1,1-dimethylethoxy)carbonyl]amino]-2-hydroxy-3-phenylpropanoate, 10-Deoxy-10-oxo-7-epidocetaxel, Docetaxel Trihydrate Imp D (EP), 4-epi-6-Oxodocetaxel, (2aR,4R,4aS,9S,11S,12S,12aR,12bS)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-Dodecahydro-4,9,11,12,12b-pentahydroxy-4a,8,13,13-tetramethyl-7,11-methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5,6-dione 12b-acetate 12-benzoate 9-ester with (2R,3S)-N-tert-butoxycarbonyl-3-phenylisoserine
InChI Formula InChI=1S/C43H51NO14/c1-22-26(55-37(51)32(48)30(24-15-11-9-12-16-24)44-38(52)58-39(3,4)5)20-43(53)35(56-36(50)25-17-13-10-14-18-25)33-41(8,34(49)31(47)29(22)40(43,6)7)27(46)19-28-42(33,21-54-28)57-23(2)45/h9-18,26-28,30,32-33,35,46,48,53H,19-21H2,1-8H3,(H,44,52)/t26-,27+,28+,30-,32+,33-,35-,41+,42-,43+/m0/s1
Solution Type Neat
SMILES CC(=O)O[C@@]12CO[C@@H]1C[C@@H](O)[C@]3(C)[C@@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](O)[C@@H](NC(=O)OC(C)(C)C)c6ccccc6)C(=C(C(=O)C3=O)C5(C)C)C
Molecular Weight (g/mol) 805.86
Quantity 25 mg
Formula Weight 805.331 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.