missing translation for 'onlineSavingsMsg'
Learn More

10-Deacetylbaccatin III, MedChemExpress

Product Code. 16441226
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441226 200 mg 200mg
16411816 500 mg 500mg
16407126 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16441226 Supplier Medchem Express Supplier No. HY16565
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 10-Deacetylbaccatin-III is an intermediate for taxol analog preparations.

10-Deacetylbaccatin-III is an intermediate for taxol analog preparations.

Specifications

Chemical Name or Material 10-Deacetylbaccatin III
CAS 32981-86-5
Color White
Purity Grade Notes Research
Molecular Formula C29H36O10
Quantity 200 mg
Solubility Information DMSO : 50 mg/mL (91.81 mM; Need ultrasonic)
SMILES CC1=C([C@@H](O)C([C@@]2(C)[C@@]3([H])[C@@](CO4)(OC(C)=O)[C@@]4([H])C[C@@H]2O)=O)C(C)(C)[C@@]([C@H]3OC(C5=CC=CC=C5)=O)(O)C[C@@H]1O
Molecular Weight (g/mol) 544.59
Formula Weight 544.59
Percent Purity 98.22%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.