missing translation for 'onlineSavingsMsg'
Learn More

10-Deacetylbaccatin III, 98%

Product Code. 15545066 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15545066 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15545066 Supplier Thermo Scientific Acros Supplier No. 294770010
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 32981-86-5 | C29H36O10 | 544.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 32981-86-5
Melting Point 221.0°C to 223.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C29H36O10
Quantity 1 g
Specific Rotation -45
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol, acetone, pyridine, chloroform
InChI Key YWLXLRUDGLRYDR-ZUKOLHLZSA-N
SMILES CC1=C2C(C(=O)C3(C(CC4C(C3C(C(C2(C)C)(CC1O)O)OC(=O)C5=CC=CC=C5)(CO4)OC(=O)C)O)C)O
Molecular Weight (g/mol) 544.6
PubChem CID 129316875
Formula Weight 544.6
Percent Purity 99%
Chemical Name or Material 10-Deacetylbaccatin III
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.