missing translation for 'onlineSavingsMsg'
Learn More

10-Deacetylbaccatin III, 95%, Thermo Scientific™

Product Code. 15467499
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467499 5 mg 5mg
15477499 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467499 Supplier Thermo Scientific Alfa Aesar Supplier No. J66228.#0

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

An intermediate in the synthesis of paclitaxel

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32981-86-5
Molecular Formula C29H36O10
Molecular Weight (g/mol) 544.597
MDL Number MFCD00132913
InChI Key YWLXLRUDGLRYDR-ZUKOLHLZSA-N
PubChem CID 129316875
SMILES CC1=C2C(C(=O)C3(C(CC4C(C3C(C(C2(C)C)(CC1O)O)OC(=O)C5=CC=CC=C5)(CO4)OC(=O)C)O)C)O

Specifications

Melting Point 229°C to 236°C
Color White
Boiling Point 717°C
Flash Point >110°C (230°F)
Quantity 5 mg
Solubility Information Soluble in chloroform; Also soluble in methanol with warming; Insoluble in water
Formula Weight 544.59
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 10-Deacetylbaccatin III
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.