missing translation for 'onlineSavingsMsg'
Learn More

10-Deacetyl Baccatin III, TRC

Product Code. 30357600
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357600 5 mg 5mg
30368274 50 mg 50mg
30351942 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357600 Supplier Toronto Research Chemicals Supplier No. TRCD1982505MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32981-86-5
Molecular Formula C29 H36 O10
Molecular Weight (g/mol) 544.59
Synonym 7,11-Methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, 12b-(acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-4,6,9,11-tetrahydroxy-4a,8,13,13-tetramethyl-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-, 7,11-Methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, 12b-(acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-4,6,9,11-tetrahydroxy-4a,8,13,13-tetramethyl-, [2aR-(2aα,4β,4aβ,6β,9α,11α,12α,12aα,12bα)]-, Tax-11-en-9-one, 5β,20-epoxy-1,2α,4,7β,10β,13α-hexahydroxy-, 4-acetate 2-benzoate (8CI), (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-(Acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-4,6,9,11-tetrahydroxy-4a,8,13,13-tetramethyl-7,11-methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, 10-Deacetylbaccatin III, 10-Desacetylbaccatin III, 10β-Deacetylbaccatin III, NSC 251677
IUPAC Name [(1S,2S,3R,4S,7R,9S,10S,12R,15S)-4-acetyloxy-1,9,12,15-tetrahydroxy-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.03,10.04,7]heptadec-13-en-2-yl] benzoate
SMILES CC(=O)O[C@@]12CO[C@@H]1C[C@H](O)[C@]3(C)[C@@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](O)C(=C([C@@H](O)C3=O)C5(C)C)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C29H36O10/c1-14-17(31)12-29(36)24(38-25(35)16-9-7-6-8-10-16)22-27(5,23(34)21(33)20(14)26(29,3)4)18(32)11-19-28(22,13-37-19)39-15(2)30/h6-10,17-19,21-22,24,31-33,36H,11-13H2,1-5H3/t17-,18-,19+,21+,22-,24-,27+,28-,29+/m0/s1
Formula Weight 544.2308
Percent Purity >95%
Chemical Name or Material 10-Deacetylbaccatin-III
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.