missing translation for 'onlineSavingsMsg'
Learn More

10-Bromodecanoic acid, 95%

Product Code. 15496758
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496758 25 g 25g
15486758 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496758 Supplier Thermo Scientific Alfa Aesar Supplier No. H60292.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50530-12-6 | C10H19BrO2 | 251.164 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Partly miscible in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50530-12-6
Molecular Formula C10H19BrO2
Molecular Weight (g/mol) 251.164
MDL Number MFCD00014388
InChI Key PGVRSPIEZYGOAD-UHFFFAOYSA-N
Synonym 10-bromo-decanoic acid, decanoic acid, 10-bromo, 10-bromanyldecanoic acid, 10-bromodecanoicacid, acmc-1aonn, 10-bromodecanoic acid., .omega.-bromodecanoic acid, ghl.pd_mitscher_leg0.481, ksc489q1r, 10-bromodecanoic acid
PubChem CID 142712
IUPAC Name 10-bromodecanoic acid
SMILES C(CCCCC(=O)O)CCCCBr

Specifications

Melting Point 38°C to 41°C
Linear Formula Br(CH2)9CO2H
Quantity 25 g
Solubility Information Partly miscible in water.
Formula Weight 251.16
Percent Purity 95%
Chemical Name or Material 10-Bromodecanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.