missing translation for 'onlineSavingsMsg'
Learn More

10-Bromo-1-decanol, 95%

Product Code. 10493355 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10493355 25 g Glass bottle 25g
10288943 1 g Glass bottle 1g
10318483 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10493355 Supplier Thermo Scientific Acros Supplier No. 384590250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53463-68-6 | C10H21BrO | 237.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53463-68-6
Molecular Formula C10H21BrO
Molecular Weight (g/mol) 237.18
MDL Number MFCD00041681
InChI Key LGZMUUBPTDRQQM-UHFFFAOYSA-N
Synonym 10-bromo-1-decanol, 10-bromodecanol, 1-decanol, 10-bromo, 10-hydroxy-1-bromo decane, 1-bromodecan-10-ol, 10-bromodecane-1-ol, 10-bromo-decan-1-ol, decamethylene bromohydrin, 10-hydroxy1-bromo decane, acmc-209l7p
PubChem CID 104507
IUPAC Name 10-bromodecan-1-ol
SMILES OCCCCCCCCCCBr

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 1.1900g/mL
Boiling Point 165°C to 169°C (10.0 mmHg)
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)10OH
Refractive Index 1.4780 to 1.4800 (20°C, 589nm)
Quantity 25 g
Specific Gravity 1.19
Formula Weight 237.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 10-Bromo-1-decanol
Show More Show Less
compliance-icons
Product Identifier
  • 10-Bromo-1-decanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.