missing translation for 'onlineSavingsMsg'
Learn More

[10-(2,5-Dihydroxy-3,4-dimethylphenyl)decyl]triphenyl-phosphonium Chloride (>90%), TRC

Product Code. 30376121
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376121 250 mg 250mg
30385310 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30376121 Supplier Toronto Research Chemicals Supplier No. TRCD453630250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [10-(2,5-Dihydroxy-3,4-dimethylphenyl)decyl]triphenyl-phosphonium Chloride
Analyte or Component Names [10-(2,5-Dihydroxy-3,4-dimethylphenyl)decyl]triphenyl-phosphonium Chloride
Shipping Condition Room Temperature
Molecular Formula C36 H44 O2 P . Cl
InChI Formula InChI=1S/C36H43O2P.ClH/c1-29-30(2)36(38)31(28-35(29)37)20-12-7-5-3-4-6-8-19-27-39(32-21-13-9-14-22-32,33-23-15-10-16-24-33)34-25-17-11-18-26-34;/h9-11,13-18,21-26,28H,3-8,12,19-20,27H2,1-2H3,(H-,37,38);1H
Solution Type Neat
SMILES [Cl-].Cc1c(O)cc(CCCCCCCCCC[P+](c2ccccc2)(c3ccccc3)c4ccccc4)c(O)c1C
IUPAC Name 10-(2,5-dihydroxy-3,4-dimethyl-phenyl)decyl-triphenyl-phosphonium;chloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 575.16
Quantity 250 mg
Formula Weight 574.2767
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.